missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CENPE Monoclonal Antibody (1H12)
GREENER_CHOICE

Código de producto. 11594343
Change view
Click to view available options
Cantidad:
50 μL
100 μL
Tamaño de la unidad:
100 microlitros
50 microlitros
Código de producto. Cantidad unitSize
11594343 100 μL 100 microlitros
15314301 50 μL 50 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11594343

Marca: Invitrogen™ MA15758

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This target has a predicted molecular weight of 312 kDa and is only suitable for Western blot if IP is performed first.

14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigmais the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to <FONT face=Symbol>g</FONT>-irradiation and other DNA-damaging agents. 14-3-3 sigmais a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5' CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CENPE
Aplicaciones Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 1H12
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.1% sodium azide
génica CENPE
N.º de referencia del gen Q02224
Alias de gen 312kDa; AU019344; BC049989; C530022J18; Cenpe; CENP-E; centromere autoantigen E; Centromere autoantigen E (312kD); centromere protein E; centromere protein E, 312kDa; centromere-associated protein E; centromeric protein E; KIF10; kinesin 10; kinesin family member 10; kinesin superfamily protein 10; Kinesin-7; Kinesin-related protein CENPE; MCPH13; Motor domain of KIF10; N-7 kinesin; PPP1R61; protein phosphatase 1, regulatory subunit 61
Símbolos de los genes CENPE
Especie del huésped Mouse
Inmunógeno Recombinant full length CENP-E protein (human) from SP2/0 cells.
Método de purificación Ion-exchange chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1062
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.