Learn More
Thermo Scientific Chemicals Celulasa, purificada cromatográficamente, T. reesei
Código de producto. p-7076619
Change view
Click to view available options
Cantidad:
250 mg
1 g
Tamaño de la unidad:
1 g
250 miligramos
15420337 250 mg 250 miligramos
15416159 1 g 1 g
2 options
Código de producto. 15420337
Marca: Thermo Scientific Chemicals J64019.MD
Solicitar formato a granel o personalizado
Cellulase has been used to degrade cello-oligosaccharides into glucose and to study the biodegradability of bioabsorbable bacterial cellulose (BBC) material. It has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. It is also applied in the biodegradation of paper products such as foolscap paper, filter paper, newspaper and office paper as well as microcrystalline cellulose. It is added to detergents to improve cleansing properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 250 mg
Formulario Liofilizado