Learn More
Thermo Scientific™ CellSensor™ CRE-bla FreeStyle™ 293F Cell Line
Descripción
The CellSensor™ CRE-bla FreeStyle™ 293F cell line contains a beta-lactamase reporter gene under control of the CRE response element stably integrated into FreeStyle™ 293F cells. To construct this cell line, the CRE -bla construct was transduced into FreeStyle™ 293F cells by lentivirus. Flow cytometry was used to isolate cells responsive to forskolin. Assay development experiments performed on this cell line included DMSO tolerance, cell number, stimulation time and substrate loading time. Functional validation of this cell line under optimized conditions included and Z' and EC50 determinations using forskolin as a primary agonist. The CellSensor™ CRE-bla FreeStyle™ 293F cell line is responsive to forskolin and can be used to build specific G-protein coupled receptor (GPCR) assays after transfection of additional genes of interest or to detect changes in cellular cAMP levels. This cell line can be adapted for high-throughput screening for agonists or antagonists with compound libraries. Candidate drugs can be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Especificaciones
Especificaciones
| Entrada del ensayo | Gen informador de beta lactamasa a base de células |
| Estado de célula | Células divisoras |
| Contenido y almacenamiento | Las líneas de células CellSensor™ se envían en hielo seco. Se debe almacenar en nitrógeno líquido inmediatamente después de su recepción, o descongelar para su uso inmediato. |
| Para utilizar con (aplicación) | GPCR, vía de señalización |
| Línea de productos | CellSensor |
| Cantidad | 1 vial |
| Entrada diana | Vía PKA, cAMP, cAMP/PKA |
| Tipo | Células en proceso de división |
| System Type | CellSensor™ |
| Druggable Target | Ruta de señalización |
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.