missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Cefotaxime
Click to view available options
Cantidad:
1 g
5 g
25 g
Tamaño de la unidad:
1 g
25 g
5 g
Descripción
Cefotaxime Sodium Salt is a third-generation cephalosporin antibiotic.
Cefotaxime, as a cephalosporin, exerts its antibacterial effects by inhibiting bacterial cell wall synthesis. It does this by binding to penicillin-binding proteins (PBPs), disrupting the final step of peptidoglycan synthesis in the bacterial cell wall. This interference leads to structural instability and eventual lysis of the bacterial cell.
Cefotaxime sodium salt is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cefotaxime.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Cefotaxime |
| CAS | 64485-93-4 |
| Cantidad | 25 g |
| Fórmula molecular | C16H16N5NaO7S2 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido