missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM7 Monoclonal Antibody (BAC2), Invitrogen™
Descripción
This antibody recognizes native human CEACAM7 protein expressed in transfected mammalian cells.
CEACAM7 is a CEA-related cell adhesion molecule 7 that belongs to the carcinoembryonic antigen (CEA) family. CEACAM7 encodes a glycosyl phosphatidyl inositol (GPI)-linked glycoprotein which is only found on the apical surface of highly differentiated epithelial cells adult colon and on a small epithelial cell population in pancreatic ducts. CEACAM7 expression is completely lost upon malignant transformation already in hyperplastic polyps and early adenomas as well as in adenocarcinomas of the colon. Like all members of the CEACAM family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by one immunoglobulin constant-like A domain.
Especificaciones
Especificaciones
| Antígeno | CEACAM7 |
| Aplicaciones | ELISA, Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | BAC2 |
| Concentración | 2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative; pH 7.2 |
| génica | CEACAM7 |
| N.º de referencia del gen | Q14002 |
| Alias de gen | BGP; BGP7; carcinoembryonic antigen CGM2; carcinoembryonic antigen gene family member 2; carcinoembryonic antigen related cell adhesion molecule 7; carcinoembryonic antigen-related cell adhesion molecule 7; CEA; CEACAM7; CGM2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?