missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CEACAM3 Monoclonal Antibody for Flow
This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Especificaciones
Especificaciones
| Antígeno | CEACAM3 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 06 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | CEACAM3 |
| N.º de referencia del gen | P40198 |
| Alias de gen | carcinoembryonic antigen CGM1; Carcinoembryonic antigen gene family (CGM1); carcinoembryonic antigen gene family 4; carcinoembryonic antigen gene family member 1; Carcinoembryonic antigen gene family member 4 / (carcinoembryonic antigene-related protein); carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen-related cell adhesion molecule 3; CD66d; CD66d antigen; CEA; cea12; CEACAM3; ceacam3 {ECO:0000312; Ceacam5; Cear; Cgm1; Cgm4; CGM4AA; EG384557; nonspecific cross-reacting antigen; pregnancy-specific glycoprotein 24; Psg24; RATCEAA; RATCGM4AA; RGD:2333}; W264; W282 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
