missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEA (CEACAM5) Monoclonal Antibody (OTI5F2), TrueMAB™, OriGene
Description
CEA (CEACAM5) Monoclonal antibody specifically detects CEA (CEACAM5) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
| Antígeno | CEA (CEACAM5) |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI5F2 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | Ceacam5 |
| N.º de referencia del gen | P06731 |
| Alias de gen | 1600029H12Rik; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CEA; CEACAM5; Meconium antigen 100; OTTHUMP00000199033; pregnancy-specific glycoprotein 30; Psg30; sCD66e; soluble CD66e |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?