missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDX2 Monoclonal Antibody (SP54)
GREENER_CHOICE

Product Code. 13461077
Change view
Click to view available options
Cantidad:
500 μL
Unit Size:
500 microlitros
Product Code. Cantidad unitSize
13461077 500 μL 500 microlitros
1 options
This item is not returnable. View return policy

Product Code. 13461077

Brand: Invitrogen™ MA516347

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is colon adenocarcinoma.

The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CDX2
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Monoclonal
Clon SP54
Concentración 0.052 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA and 0.1% sodium azide; pH 7.2
génica CDX2
N.º de referencia del gen Q99626
Alias de gen caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor
Símbolos de los genes CDX2
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to N-terminus of human CDX2.
Método de purificación Protein A/G
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1045
Especies diana Human
Contenido y almacenamiento 4°C, do not freeze
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.