missing translation for 'onlineSavingsMsg'
Learn More

CDKN2C Monoclonal Antibody (OTI5G1), TrueMAB™, OriGene

Product Code. 17549532
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
Product Code. Cantidad unitSize
17549532 100 μL 100 microlitros
17569532 30 μL 30 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17549532

Brand: Origene Technologies TA801230

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to interact with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. Ectopic expression of this gene was shown to suppress the growth of human cells in a manner that appears to correlate with the presence of a wild-type RB1 function. Studies in the knockout mice suggested the roles of this gene in regulating spermatogenesis, as well as in suppressing tumorigenesis. Two alternatively spliced transcript variants of this gene, which encode an identical protein, have been reported.

Specifications

Antígeno CDKN2C
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI5G1
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica CDKN2C
N.º de referencia del gen P42773
Alias de gen INK4C, p18, p18-INK4C
Símbolos de los genes CDKN2C
Especie del huésped Mouse
Inmunógeno Full length human recombit protein of human CDKN2C produced in E.coli.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1031
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.