missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA511540
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with chicken and mouse based on sequence homology.
This gene is a type II classical cadherin from the cadherin superfamily. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Cadherins mediate cell-cell binding in a homophilic manner, contributing to the sorting of heterogeneous cell types and the maintenance of orderly structures. Alternative splicing in the 5' UTR of this gene yields variant transcripts encoding the same protein.
Especificaciones
| CDH7 | |
| Polyclonal | |
| Unconjugated | |
| CDH7 | |
| 9330156F07Rik; Cad7; Cadherin; cadherin 7; cadherin 7, type 2; Cadherin7; cadherin-7; Cdh7; CDH7L1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1005 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9ULB5 | |
| CDH7 | |
| KLH conjugated synthetic peptide between 56-84 amino acids from the N-terminal region of human CDH7. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido