Learn More
DiscoverX CDDO Methyl Ester
Código de producto. 17392469
Encompass
Change view
Click to view available options
:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto.
17392469 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 17392469 Proveedor DiscoverX N.º de proveedor 921147
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
CDDO Methyl Ester is an agonist for the KEAP1-Nrf2 pathway. Provided as 5mg of dry powder.
CDDO Methyl Ester (bardoxolone methyl) is a semi-synthetic triterpenoid developed from the oleanolic acid scaffold that induces apoptosis and differentiation in tumor cells. Agonist of the KEAP1-Nrf2 pathway, thus inducing expression of antioxidant proteins and protection against oxidative damage from injury and inflammation. Also an antagonist of the NF-kB pro-inflammatory transcription factor and PPARG. Underwent clinical trials for type 2 diabetes treatment, due to its potent antidiabetic activity, but was confirmed to enhance cardiac disease during a phase 2 trial in October 2012. Provided as 5mg of dry powder.
Especificaciones
Cantidad 5 mg