Learn More
CD97 Mouse anti-Human, Unconjugated, Clone: 3F10C2, Proteintech
Código de producto. 16889305 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16889305 20 μL 20 microlitros
16879305 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16889305 Proveedor Proteintech N.º de proveedor 669721IG20UL
Mouse Monoclonal Antibody
The CD97 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD97. This antibody recognizes human, mouse antigen. The CD97 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Especificaciones
Antígeno CD97
Aplicaciones Western Blot, Flow Cytometry, ELISA
Clasificación Monoclonal
Clon 3F10C2
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 976
Especies diana Human, Mouse
Isotype IgG2a