missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD93 Mouse anti-Human, Oligo, Clone: R139, BD™ AbSeq ™
Descripción
CD93 Monoclonal antibody specifically detects CD93 in Human samples. It is validated for Single Cell 3' Sequencing
Especificaciones
Especificaciones
| Antígeno | CD93 |
| Aplicaciones | Single Cell 3' Sequencing |
| Clasificación | Monoclonal |
| Clon | Clone R139 |
| Conjugado | Oligo |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Alias de gen | C1QR1; C1qRP; C1qR(P); C1q/MBL/SPA Receptor; MXRA4; GR11; Dj737e23.1 |
| Especie del huésped | Mouse |
| Inmunógeno | Clq-CLF-binding proteins |
| Cantidad | 25 Tests |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?