missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD91 Monoclonal Antibody (A2MR-a2), NovaFluor™ Red 725, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
160.00€ - 355.00€
Especificaciones
| Antígeno | CD91 |
|---|---|
| Clon | A2MR-a2 |
| Concentración | 0.05 μg/Test |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
Descripción
The protein encoded by this gene is an endocytic receptor involved in several cellular processes, including intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. In addition, the encoded protein is necessary for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of this gene decreases with age and has been found to be lower than controls in brain tissue from Alzheimer patients.Especificaciones
| CD91 | |
| 0.05 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A2MR; AI316852; alpha 2-macroglobulin receptor; alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; APR; b2b1554Clo; CD91; FLJ16451; IGFBP3R; LDL receptor related protein 1; lipoprotein receptor-related protein; low density lipoprotein receptor-related protein 1; low density lipoprotein-related protein 1 (alpha-2-macroglobulin receptor); Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; Lrp; Lrp1; LRP-1; LRP1A; LRP-515; LRP-85; LRPICD; MGC88725; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; TGFBR5; type V tgf-beta receptor | |
| LRP1 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| LRP1 |
| A2MR-a2 | |
| Flow Cytometry | |
| NovaFluor Red 725 | |
| Mouse | |
| Human | |
| Q07954 | |
| 4035 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto