missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Thy1 Antibody (G7), Biotin, Novus Biologicals™
Rat Monoclonal Antibody has been used in 5 publications
Marca: Novus Biologicals NBP1-28033-500UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay.
Especificaciones
| CD90/Thy1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry < 1 μg/106 cells, Immunohistochemistry, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, In vitro assay | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| Rat | |
| 500 μg | |
| 7070 | |
| Mouse | |
| Purified | |
| Mesenchymal Cell Marker |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), In vitro Assay | |
| G7 | |
| Biotin | |
| PBS | |
| THY1 | |
| Mouse T cell hybridoma C6/G8 | |
| Hematopoietic Stem Cell Markers, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| Mouse CD90 (Thy-1.1 & Thy-1.2 alloantigens/Thy-1 epitope region A); Mr 28-32 kDa | |
| Store at 4°C. Do not freeze. | |
| IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction