missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 (Thy-1) Mouse, anti-Human, PE, Clone: 5E10, STEMCELL Technologies
Descripción
- The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
- CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
- It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
- CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
- This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Especificaciones
Especificaciones
| Antígeno | CD90 (Thy-1) |
| Aplicaciones | Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 5E10 |
| Conjugado | PE |
| Especie del huésped | Mouse |
| Cantidad | 100 Tests |
| Primario o secundario | Primary |
| Especies diana | Baboon, Cynomolgus, Rhesus Monkey, Pig, Human |
| Isotype | IgG1 κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?