Learn More
CD90 Mouse anti-Human, Porcine, Rat, Clone: 2D7D11, Proteintech
Código de producto. 16848945 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16848945 20 μL 20 microlitros
16838945 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16848945 Proveedor Proteintech N.º de proveedor 667661IG20UL
Mouse Monoclonal Antibody
THY1 encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins. The encoded protein is involved in cell adhesion and cell communication in numerous cell types, but particularly in cells of the immune and nervous systems. The encoded protein is widely used as a marker for hematopoietic stem cells. This gene may function as a tumor suppressor in nasopharyngeal carcinoma. Alternative splicing results in multiple transcript variants.
Especificaciones
Antígeno CD90
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 2D7D11
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica THY1
N.º de referencia del gen P01830, P04216
Alias de gen CD90, CDw90, FLJ33325, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1
Símbolos de los genes THY1
Especie del huésped Mouse
Inmunógeno CD90 Fusion Protein Ag25603
Método de purificación Protein A
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100271931, 24832, 7070
Especies diana Human, Rat, Pig
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Mostrar más Mostrar menos
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.