missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen™ M028T03R04-A
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD90.2 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90.2 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90.2 is an 18 kDa plasma membrane antigen expressed by hematopoietic cells and neurons in all species studied, highly expressed in connective tissue, various fibroblast and stromal cell lines and lastly, by tumor endothelial cell lines. In the mouse, CD90.2 is expressed on all thymocytes and peripheral T lymphocytes.
Especificaciones
| CD90.2 (Thy-1.2) | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01831 | |
| Thy1 | |
| Affinity chromatography | |
| RUO | |
| 21838 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG2a κ |
| Flow Cytometry | |
| 53-2.1 | |
| NovaFluor Red 710 | |
| Thy1 | |
| CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido