missing translation for 'onlineSavingsMsg'
Learn More

CD8b.2 Rat anti-Mouse, BV750, Clone: 53-5.8, BD Biosciences™

Código de producto. 17299424
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17299424 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17299424 Proveedor BD Biosciences N.º de proveedor 747512

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

CD8b.2 Monoclonal antibody specifically detects CD8b.2 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD8b.2
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 53-5.8
Conjugado Brilliant Violet 750
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Alias de gen Ly-3.2; Lyt-3.2; CD8b.2
Especie del huésped Rat
Inmunógeno Mouse thymus or spleen
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Mouse
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody.
Isotype IgG1 κ
Mostrar más Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.