missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Mouse, anti-Human, FITC, Clone: RPA-T8, STEMCELL Technologies
Descripción
- The RPA-T8 antibody reacts with CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
- CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
- CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
- It functions to strengthen the association between the TCR and MHC I-antigen complex
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD8+ T Cell Isolation Kit (Catalog #19663), EasySep™ Human CD8+ T Cell Enrichment Kit (Catalog #19053), EasySep™ Human CD8 Positive Selection Kit II (Catalog #17853), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851), EasySep™ Human CD2 Positive Selection Kit II (Catalog #17883), and EasySep™ HLA T Cell Enrichment Kit (Catalog #19051HLA)
Especificaciones
Especificaciones
| Antígeno | CD8a |
| Aplicaciones | CyTOF, Flow Cytometry, Functional Assay, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | RPA-T8 |
| Conjugado | FITC |
| Especie del huésped | Mouse |
| Cantidad | 25 Tests |
| Primario o secundario | Primary |
| Especies diana | Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human |
| Isotype | IgG1 κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?