Learn More
CD86 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16831584 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16831584 150 μL 150 microlitros
16841584 20 μL 20 microlitros
2 options
Código de producto. 16831584
Marca: Proteintech 133951AP150UL
Rabbit Polyclonal Antibody
This antibody detects a band of approximately 70 kDa, consistent with glycosylated form of CD86.
CD86 is one of two ligands (the other CD80) for CTLA4 and CD28. CD86 acts as costimulatory molecule in eliciting T-cell help during antigen presentation. Antigen presentation in the absence of sufficient co-stimulation involving CD86/CD80 can induce tolerance. CD80 appears to play a role distinct from CD80 in T helper cell differentiation. CD86 is a type I membrane protein that is a member of the immunoglobulin superfamily. The CD86 protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of CD86 with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of CD86 with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms of CD86. Additional transcript variants have been described for CD86, but their full-length sequences have not been determined. Diseases associated with CD86 dysfunction include gallbladder squamous cell carcinoma and myocarditis.
Especificaciones
Antígeno CD86
Aplicaciones Flow Cytometry, Western Blot
Clasificación Polyclonal
Concentración 0.19 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CD86
N.º de referencia del gen P42081
Alias de gen Activation B7 2 antigen, B7 2, B70, BU63, CD28LG2, CD86, CD86 molecule, FUN 1, LAB72
Símbolos de los genes CD86
Especie del huésped Rabbit
Inmunógeno CD86 Fusion Protein Ag4063
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 942
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG