Learn More
CD86 Mouse anti-Rat, FITC, Clone: 24F, BD
Descripción
The 24F antibody reacts with CD86 (B7-2), a member of the Ig superfamily of transmembrane proteins. CD86, a ligand for CD28 and CD152 (CTLA-4), is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. CD86 is predominantly expressed on antigen-presenting cells. It can be upregulated on splenic B cells by in vitro polyclonal activation, such as LPS treatment. 24F mAb is reported to block the costimulatory function of rat CD86 and to immunoprecipitate CD86 from Lewis-S1 cell lysates.HTLV-1-transformed rat T-cell line Lewis-S1
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Especificaciones
Especificaciones
| Antígeno | CD86 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 24F |
| Concentración | 0.5mg/mL |
| Conjugado | FITC |
| Descripción | B7-2 |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.