Learn More
CD83 Mouse anti-Human, PE, Clone: HB15E, BD
Descripción
Reacts with a 45kDa transmembrane protein member of the Ig superfamily. CD83 is composed of a single V-type Ig extracellular domain with a C-terminal cytoplasmic tail. CD83 is found mainly on follicular dendritic cells, circulating dendritic cells, interdigitating dendritic cells in lymphoid tissues, in vitro-generated dendritic cells and thymic dendritic cells. However, its expression is not restricted to dendritic cells. CD83 is also expressed on some germinal center B cells and some lymphoblastoid cell lines. Although its function is not known, it may play a role in cell-cell interaction during antigen presentation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: HB15e
Isotype: IgG1, κ
Species Reactivity: Human
Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | CD83 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | HB15E |
| Conjugado | PE |
| Descripción | BL11; HB15; B-cell activation protein |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human CD83 transfected COS cells |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.