missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD81 Monoclonal Antibody (Eat2), FITC

Código de producto. 15384211
Change view
Click to view available options
Cantidad:
100 μg
300 μg
Tamaño de la unidad:
100 microgramos
300 microgramos
Código de producto. Cantidad unitSize
15384211 300 μg 300 microgramos
15374211 100 μg 100 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15384211

Marca: Invitrogen™ MA517940

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Armenian Hamster Monoclonal Antibody

CD81 Monoclonal Antibody for Flow

CD81 (TAPA-1, target of anti-proliferative antibody-1) is a member of the tetraspanin family, is expressed on virtually all nucleated cells, but above all on germinal center B cells. CD81 forms complexes with other tetraspanin proteins, integrins, coreceptors, MHC class I and II molecules, and influences adhesion, morphology, activation, proliferation and differentiation of B, T cells. In muscles, CD81 promotes cell fusion and myotube maintenance. CD81 has been also identified as a receptor for the hepatitis C virus. Like members of the tetraspanin family that include CD9, CD37, CD53, CD63, and CD82, CD81 is a cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD81 is a cell surface glycoprotein that is known to complex with integrins. CD81 appears to promote muscle cell fusion and support myotube maintenance. CD81 associates with CD19, CD21, Leu 13, and integrins on cell membrane and acts as a receptor for the envelope protein E2 of chronic hepatitis C virus. Antibodies to CD81 have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. CD81 is also localized in the tumor-suppressor gene region and is a candidate gene for malignancies.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD81
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon Eat2
Concentración 0.1 mg/mL
Conjugado FITC
Formulación PBS with 4-5mg/mL BSA and 0.02% sodium azide
génica CD81
N.º de referencia del gen P35762, Q62745
Alias de gen 26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28
Símbolos de los genes CD81
Especie del huésped Armenian Hamster
Inmunógeno CD81 + mouse B lymphoma 38C13.
Método de purificación Protein G
Cantidad 300 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12520, 25621
Especies diana Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.