missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD79a Monoclonal Antibody (HM47), Alexa Fluor™ 647

Descripción
CD79a Monoclonal Antibody for IHC (P), Flow
CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
Especificaciones
Especificaciones
| Antígeno | CD79a |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | HM47 |
| Concentración | 0.05 mg/mL |
| Conjugado | Alexa Fluor 647 |
| Formulación | PBS with 1% BSA and 0.0325% sodium azide; pH 7.4 |
| génica | CD79A |
| N.º de referencia del gen | P11912 |
| Alias de gen | B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?