Learn More
CD71 Rabbit, anti-Human, Polyclonal, Proteintech
Código de producto. 18035015 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
18035015 20 μL 20 microlitros
18084505 150 μL 150 microlitros
2 options
Código de producto. 18035015
Marca: Proteintech 305741AP20UL
Rabbit Polyclonal Antibody
The CD71 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD71. This antibody recognizes human antigen. The CD71 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Especificaciones
Antígeno CD71
Aplicaciones Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
Concentración 260 μg/mL
Símbolos de los genes CD71, p90, sTfR, T9, TFR, TFR1, TFRC, TR, Transferrin receptor protein 1, TRFR
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 7037
Especies diana Human
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG