Learn More
CD70 Mouse anti-Human, PE, Clone: KI-24, BD
Código de producto. 15801489
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
cada uno
15801489 25 Tests cada uno
15888468 100 Tests cada uno
2 options
Código de producto. 15801489
Marca: BD Biosciences 561935
Mouse Monoclonal Antibody
The Ki-24 monoclonal antibody specifically binds to human CD70. CD70 is a type II transmembrane glycoprotein and member of the TNF Superfamily. CD70 is also known as Tumor necrosis factor ligand superfamily member 7 (TNFSF7), CD27 ligand (CD27-L, CD27L, CD27LG), and KI-24 antigen. The CD70 antigen immunoprecipitates as five bands (50, 70, 90, 100 and 160 kDa) under non-reducing conditions. CD70 is strongly expressed on Reed-Sternberg cells, some activated T or B cells and Epstein Barr Virus (EBV)-positive lymphoblastoid cell lines. CD70 plays roles in the activation, proliferation and differentiation of B cells and T cells including the enhanced production of cytotoxic T cells.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD70
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon Ki-24
Conjugado PE
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Método de purificación Affinity Purified
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG3 κ
Mostrar más Mostrar menos
For Research Use Only.