missing translation for 'onlineSavingsMsg'
Learn More

CD69 Hamster anti-Mouse, PE-Cy7, Clone: H1.2F3, BD

Hamster Monoclonal Antibody

107.00€ - 286.00€

Especificaciones

Antígeno CD69
Clon H1.2F3
Concentración 0.2mg/mL
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15851479
Ver documentos
BD Biosciences
561930
25 μg
107.00€
cada uno
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
15846888
Ver documentos
BD Biosciences
552879
0.1 mg
286.00€
cada uno
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

The H1.2F3 monoclonal antibody specifically binds to CD69 (Very Early Activation antigen), an 85 kDa disulfide-linked homodimer of differentially glycosylated subunits. CD69 is a C-type lectin, most closely related to the NKR-P1 and Ly-49 NK cell-activation molecules. Its expression is rapidly induced upon activation of lymphocytes (T, B, NK, and NK-T cells), neutrophils, and macrophages. CD69 is expressed also on thymocytes that are undergoing positive selection; its role in that process is unclear. H1.2F3 mAb augments PMA-induced T-cell stimulation and IFN-γ-induced macrophage stimulation. IL-2-activated NK cells express CD69, and H1.2F3 mAb induces redirected lysis of FcR-bearing target cells by NK cells.

Especificaciones

Especificaciones

CD69
0.2mg/mL
Monoclonal
Hamster
Mouse
Mouse Dendritic Epidermal T Cell Line Y245
Affinity Purified
VEA; Very Early Activation Antigen; AIM; Activation Induced Molecule
H1.2F3
Flow Cytometry
PE-Cyanine7
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.