Learn More
CD68 Monoclonal Antibody (OTI13H2), TrueMAB™, OriGene
Product Code. 17531853
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
17531853 100 μL 100 microlitros
17551853 30 μL 30 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17531853 Supplier Origene Technologies Supplier No. TA802981
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
This item is not returnable. View return policy
Mouse Monoclonal Antibody
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predomitly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human maligcies, including several Acute Myeloid Leukemia studies.
Specifications
Antígeno CD68
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI13H2
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica CD68
N.º de referencia del gen P34810
Alias de gen GP110, LAMP4, SCARD1
Símbolos de los genes CD68
Especie del huésped Mouse
Inmunógeno Human recombit protein fragment corresponding to amino acids 22-319 of human CD68 produced in SF9 cell.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 968
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1