missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD66b Mouse, anti-Human, PE, Clone: G10F5, STEMCELL Technologies
Descripción
- The G10F5 antibody reacts with CD66b (previously denoted CD67), an ∼95 kDa single-chain glycosylphosphatidylinositol (GPI)-linked glycoprotein expressed by neutrophilic and eosinophilic granulocytes
- CD66b is a member of the carcinoembryonic antigen (CEA)-like subfamily of proteins within the immunoglobulin superfamily and possesses two Ig-like C2-type domains and one Ig-like V-type domain
- In neutrophils, CD66b is localized within granules in the cytoplasm as well as being expressed on the plasma membrane
- Surface expression levels increase upon granulocyte activation, an effect induced by stimulators such as phorbol myristate acetate (PMA), calcium ionophore, and N-formylmethionyl-leucyl-phenylalanine (FMLP)
- CD66b appears to play a role as a signaling receptor involved in cell adhesion, phagocytosis, and chemotaxis by regulating granulocyte activation and adhesion to several proteins, including CD66c, galectin-3, fibronectin, and E-selectin
Especificaciones
Especificaciones
| Antígeno | CD66b |
| Aplicaciones | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | G10F5 |
| Conjugado | PE |
| Especie del huésped | Mouse |
| Cantidad | 25 Tests |
| Primario o secundario | Primary |
| Especies diana | Chimpanzee, Human |
| Isotype | IgM κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?