Learn More
Invitrogen™ CD62L Recombinant Rabbit Monoclonal Antibody (414), APC

Descripción
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Mouse L-Selectin (CD62L) (Met1-Asn332) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
Especificaciones
Especificaciones
| Antígeno | CD62L |
| Aplicaciones | Flow Cytometry |
| Clasificación | Recombinant Monoclonal |
| Clon | 414 |
| Concentración | 100 μg/mL |
| Conjugado | APC |
| Formulación | PBS with 0.5% BSA and 0.09% sodium azide |
| génica | SELL |
| N.º de referencia del gen | P18337 |
| Alias de gen | A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.