Learn More
CD54 (ICAM-1) Rat, anti-Mouse, Unconjugated, Clone: YN1/1.7.4, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60151
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17129191
  • 353.70€ / 500 microgramos
Descripción
  • The YN1/1.7.4 antibody reacts with CD54, a 90 - 110 kDa type I transmembrane glycoprotein that is a member of the immunoglobulin (Ig) superfamily
  • CD54 consists of five Ig-like C2 type domains and has two isoforms generated from alternative splicing
  • As an important intercellular adhesion molecule, CD54 is involved in inflammatory reactions, antigen-specific immune responses, lymphocyte trafficking, and T cell signal transduction events
  • CD54 mediates important cell-cell interactions and adhesion reactions by binding to its major ligands, LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18)
  • CD54 may also act as a receptor for rhinoviruses and some coxsackieviruses
  • CD54 is expressed on lymphocytes, monocytes, macrophages, dendritic cells, endothelial cells, and high endothelial venules
  • Expression of CD54 is upregulated on activated lymphocytes and also, following stimulation with inflammatory mediators such as cytokines, LPS, and IFN-γ
Especificaciones
Especificaciones
CD54 (ICAM-1)
Monoclonal
Unconjugated
500 μg
Mouse
Endothelial Cells
Immunoblot, Flow Cytometry, Immunoprecipitation
YN1/1.7.4
Rat
Primary
IgG2b κ
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.