missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD5 Monoclonal Antibody (CG16)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA170072
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA1-70072 detects CD5 in mouse samples. MA1-70072 has been successfully used in flow cytometry procedures. The MA1-70072 immunogen is C3H.CE - Ly 1.2 : DS.
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter's Syndrome.
Especificaciones
| CD5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P13379 | |
| Cd5 | |
| C3H.CE - Ly 1.2: DS. | |
| 250 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Cytotoxicity Assay, Flow Cytometry | |
| CG16 | |
| Unconjugated | |
| Cd5 | |
| alanyl (membrane) aminopeptidase; alanyl aminopeptidase; alanyl aminopeptidase, membrane; aminopeptidase M; Aminopeptidase N; ANPEP; AP-M; APN; AP-N; CD antigen CD5; CD13; CD28; CD28 antigen (Tp44); CD28 molecule; Cd5; CD5 antigen; CD5 antigen (p56 62); CD5 antigen (p56-62); CD5 antigen p56-62; CD5 molecule; CD5 protein; CD74 antigen, invariant polypeptide of major; cell surface protein; costimulatory molecule B7 receptor CD28; fCD5; GP150; hAPN; LAP1; LEU1; Ly-1; Ly12; Ly-12; LyA; Ly-A; Lymphocyte antigen 1; Lymphocyte antigen CD5; lymphocyte antigen T1/Leu-1; Lyt-1; membrane alanyl aminopeptidase; microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13; P150; Pan T cell; PEPN; T1; T-cell costimulatory molecule CD28; T-cell surface glycoprotein CD5; T-cell-specific surface glycoprotein CD28; unnamed protein product | |
| Mouse | |
| Protein G | |
| RUO | |
| 12507 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido