missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD49a (Integrin alpha 1) Recombinant Rabbit Monoclonal Antibody (7T5B6)

Código de producto. 30131401
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
30131401 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30131401

Marca: Invitrogen™ MA555280

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: FNVDVKNSMT FSGPVEDMFG YTVQQYENEE GKWVLIGSPL VGQPKNRTGD VYKCPVGRGE SLPCVKLDLP VNTSIPNVTE VKENMTFGST LVTNPNGGFL ACGPLYAYRC GHLHYTTGIC SDVSPTFQVV NSIAPVQECS TQLDIVIVLD GSNSIYPWDS VTAFLNDLLE RMDIGPKQTQ VGIVQYGENV THEFNLNKYS STEEVLVAAK KIVQRGGRQT MTALGIDTAR KEAFTEARGA RRGVKKVMVI VTDGESHDNH RLKKVIQDCE DENIQRFSIA ILGSYNRGNL STEKFVEEIK SIASEPTEKH FFNVSDELAL VTIVKTLGER IFALEATADQ SAASFEMEMS QTGFSAHYSQ DWVMLGAVGA YDWNGTVVMQ KASQIIIPRN TTFNVESTKK NEPLASYLGY TVNSATASSG DVLYIAGQPR YNHTGQVIIY RMEDGNIKIL QTLSGEQIGS YFGSILTTTD IDKDSNTDIL LVGAPMYMGT EKEEQGKVYV YALNQTRFEY QMSLEPIKQT CCSSRQHNSC TTENKNEPCG ARFGTAIAAV KDLNLDGFND IVIGAPLEDD HGGAVYIYHG SGKTIRKEYA QRIPSGGDGK TLKFFGQSIH GEMDLNGDGL TDVTIGGLGG AALFWSRDVA VVKVTMNFEP NKVNIQKKNC HMEGKETVCI NATVCFDVKL KSKEDTIYEA DLQYRVTLDS LRQISRSFFS GTQERKVQRN ITVRKSECTK HSFYMLDKHD FQDSVRITLD FNLTDPENGP VLDDSLPNSV HEYIPFAKDC GNKEKCISDL SLHVATTEKD LLIVRSQNDK FNVSLTVKNT KDSAYNTRTI VHYSPNLVFS GIEAIQKDSC ESNHNITCKV GYPFLRRGEM VTFKILFQFN TSYLMENVTI YLSATSDSEE PPETLSDNVV NISIPVKYEV GLQFYSSASE YHISIAANET VPEVINSTED IGNEINIFYL IRKSGSFPMP ELKLSISFPN MTSNGYPVLY PTGLSSSENA NCRPHIFEDP FSINSGKKMT TSTDHLKRGT ILDCNTCKFA TITCNLTSSD ISQVNVSLIL WKPTFIKSYF SSLNLTIRGE LRSENASLVL SSSNQKRELA IQISKDGLPG RVP.

Integrin alpha 1 (ITGA1) chain associates with the beta 1 (ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD49a (Integrin alpha 1)
Aplicaciones ELISA, Western Blot
Clasificación Recombinant Monoclonal
Clon 7T5B6
Concentración 0.72 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3
génica Itga1
N.º de referencia del gen P56199
Alias de gen CD49 antigen-like family member A; CD49a; E130012M19Rik; integrin alpha 1; integrin alpha-1; integrin subunit alpha 1; integrin, alpha 1; Itga1; Laminin and collagen receptor; very late activation protein 1; Vla1; VLA-1
Símbolos de los genes Itga1
Especie del huésped Rabbit
Inmunógeno Recombinant Protein (or fragment).
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3672
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.