missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD47 Monoclonal Antibody (B6H12.2)
GREENER_CHOICE

Código de producto. 11364533
Change view
Click to view available options
Cantidad:
500 μL
Tamaño de la unidad:
500 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11364533 500 μL 500 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11364533 Proveedor Invitrogen™ N.º de proveedor MA511895

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA5-11895 targets CD47 in WB, FACS, IF, and IP applications and shows reactivity with Human samples. The MA5-11895 immunogen is intact CD47 purified from placenta.

CD47, known as integrin-associated protein (IAP), is a glycosylated transmembrane glycoprotein with five domains, expressed widely across hematopoietic cells like T and B cells, monocytes, platelets, and erythrocytes, as well as non-hematopoietic cells. It interacts with integrins such as CD51/CD61 and CD41/CD61, and serves as a receptor for thrombospondin, mediating bi-directional signaling that affects neural synaptic activity and macrophage phagocytosis. CD47 acts as a ligand for CD172a (SIRP alpha), an inhibitory receptor on macrophages, preventing phagocytosis of CD47-positive cells. This interaction influences cell migration, B cell adhesion, T cell activation, and neuronal development, particularly in synapse-rich brain and retina regions. It also modulates chondrocyte responses to mechanical signals. T cell expression of CD47 can lead to activation or apoptosis in the presence of thrombospondin. Monoclonal antibody stimulation of CD47 has been shown to induce CD4+CD25- suppressive activity and increase Foxp3 expression. CD47's role in membrane transport, signal transduction, and its broad tissue distribution highlight its significance in various physiological processes.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD47
Aplicaciones Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon B6H12.2
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4
génica CD47
N.º de referencia del gen Q08722
Alias de gen 9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47
Símbolos de los genes CD47
Especie del huésped Mouse
Inmunógeno Intact CD47 purified from placenta.
Método de purificación Protein A/G
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 961
Especies diana Human
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.