missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD46 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15308034
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
400 microlitros
Código de producto. Cantidad unitSize
15308034 400 μL 400 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15308034

Marca: Invitrogen™ PA535311

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

CD46 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow

CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD46
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.48 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica CD46
N.º de referencia del gen P15529
Alias de gen AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antigen
Símbolos de los genes CD46
Especie del huésped Rabbit
Inmunógeno KLH conjugated synthetic peptide between 317-343 amino acids from the C-terminal region of human CD46.
Método de purificación Antigen affinity chromatography, Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4179
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.