missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD46 Monoclonal Antibody (MEM-258), PE
GREENER_CHOICE

Código de producto. 11529202
Change view
Click to view available options
Cantidad:
100 Tests
Tamaño de la unidad:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11529202 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11529202 Proveedor Invitrogen™ N.º de proveedor MA119649

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on SCR4 (the membrane-proximal SCR) domain of CD46 (Membrane cofactor protein). CD46 is 56-66 kDa dimeric transmembrane protein expressed on T and B lymphocytes, platelets, monocytes, granulocytes, endothelial cells, epithelial cells and fibroblast; it is negative on erythrocytes.

CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD46
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon MEM-258
Conjugado PE
Formulación PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
génica CD46
N.º de referencia del gen P15529, Q6VE48
Alias de gen AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antigen
Símbolos de los genes CD46
Especie del huésped Mouse
Inmunógeno T cell line HPB-ALL.
Método de purificación Size-exclusion chromatography
Cantidad 100 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 280851, 4179
Especies diana Human, Bovine
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.