Learn More
CD45R (B220) Rat, anti-Mouse, Unconjugated, Clone: RA3-6B2, STEMCELL Technologies
Código de producto. 17107591
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μg
500 μg
Tamaño de la unidad:
100 microgramos
500 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17107591 100 μg 100 microgramos
17168501 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17107591 Proveedor Stemcell Technologies N.º de proveedor 1000422
Rat Monoclonal Antibody
  • The RA3-6B2 antibody reacts with an epitope on the extracellular domain of the exon A‐restricted isoform of mouse CD45
  • CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
  • The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains (one of which is enzymatically active) that are involved in modulating the function of Src kinases such as Lck and Fyn
  • Several isoforms of CD45 have been identified, the expression of which differs according to cell type and functional status
  • Alternative splicing of three exons encoding the extracellular RA, RB, and RC polypeptide sequences gives rise to up to 8 isoforms with molecular masses in the range of 200 - 220 kDa
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse B Cell Isolation Kit (Catalog #19854)
Especificaciones
Antígeno CD45R (B220)
Aplicaciones Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon RA3-6B2
Conjugado Unconjugated
Especie del huésped Rat
Cantidad 100 μg
Primario o secundario Primary
Especies diana Feline, Human, Mouse
Tipo de célula B Cells, Leukocytes, Lymphocytes