missing translation for 'onlineSavingsMsg'
Learn More

CD45R/B220 Rat anti-Mouse, PE-Cy5, Clone: RA3-6B2, BD

Código de producto. p-7092606
Change view
Click to view available options
Cantidad:
0.1 mg
25 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15841429 25 μg cada uno
15806998 0.1 mg cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15841429

Marca: BD Biosciences 561879

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

The rat anti-mouse CD45R antibody (clone RA3-6B2) has been reported to react with an epitope on the extracellular domain of the transmembrane CD45 glycoprotein which is dependent upon the expression of exon A and specific carbohydrate residues. It is expressed on B lymphocytes at all stages from pro-B through mature and activated B cell, but it is decreased on plasma cells and a subset of memory B cells. The levels of CD45R expression on the B-cell lineage appear to be developmentally regulated. It is also reportedly found on the abnormal T cells involved in the lymphadenopathy of lpr/lpr and gld/gld mutant mice, on lytically active subsets of lymphokine-activated killer cells (NK cells and non-MHC-restricted CTL), on apoptotic T lymphocytes of mice injected with bacterial superantigen, on a population of NK-cell precursors in the bone marrow, and on B-lymphocyte, T-lymphocyte, and macrophage progenitors in fetal liver. The CD45R antigen has been reported not to be on hematopoietic stem cells, naive T lymphocytes, or MHC-restricted CTL. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. CD45R is commonly used as a pan B-cell marker; however, CD19 expression, detectable by the rat anti-mouse CD19 antibody (clone 1D3), is reported to be more restricted to the B-cell lineage. The rat anti-mouse CD45R antibody (clone RA3-6B2) has been reported to enhance isotype switching during in vitro B-cell responses and to inhibit in vivo B-cell responses. Cross-reaction of the RA3-6B2 clone with activated human T lymphocytes has also been reportedly observed.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD45R
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon RA3-6B2
Concentración 0.2mg/mL
Conjugado PE-Cyanine5
Descripción B220; Ly-5; CD45R; LCA; Ptprc; Protein tyrosine phosphatase receptor type C
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Rat
Inmunógeno Mouse Abelson Leukemia Virus-Induced pre-B tumor cells
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human, Mouse
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.