Learn More
CD45.1 Mouse anti-Mouse, FITC, Clone: A20, BD
Mouse Monoclonal Antibody
140.37€ - 315.00€
Especificaciones
| Antígeno | CD45.1 |
|---|---|
| Clon | A20 |
| Concentración | 0.5mg/mL |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15861419
|
BD Biosciences
561871 |
0.1 mg |
141.00€ 140.37€ / cada uno Ahorro 0.63€ 0.45% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
15817388
|
BD Biosciences
553775 |
0.5 mg |
315.00€
cada uno |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
The A20 monoclonal antibody specifically binds to CD45 (Leukocyte Common Antigen) on all leukocytes of mouse strains expressing the CD45.1 alloantigen (eg, RIII, SJL/J, STS/A, DA). This alloantigen was originally named Ly-5.2, and this was the designation at the time that the antibody was characterized. The designation was later changed from Ly-5.2 to Ly-5.1 to conform with the convention that the .2 alloantigen designations be assigned to the C57BL/6 strain. mAb A20 has been reported not to react with leukocytes of most other mouse strains, which express the CD45.2 alloantigen. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C,respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. The A20 antibody has been reported to inhibit some responses of B cells, from mice expressing the CD45.1 alloantigen, to antigens and LPS.
Especificaciones
| CD45.1 | |
| 0.5mg/mL | |
| Monoclonal | |
| Mouse | |
| Mouse | |
| SJL mouse thymocytes and splenocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| A20 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| Ly-5; Lyt4; CD45R; LCA; Ptprc; Protein tyrosine phosphatase receptor type C |
For Research Use Only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.