missing translation for 'onlineSavingsMsg'
Learn More

CD44 Mouse anti-Human, Porcine, Clone: 6F4H2, Proteintech

Código de producto. p-7213391 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
16827245 20 μL 20 microlitros
16817245 150 μL 150 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16827245

Marca: Proteintech 602241IG20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

CD44 cell surface antigen is a 100 kDa type 1 transmembrane glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predomit CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential. CD44 is expressed by hematopoietic, non-hematopoietic cells, epithelial tissues, and to filopodia in cultured keratinocytes. Further, bone marrow myeloid cells and memory T cells express CD44 at high levels, and peripheral B and T cells can upregulate the expression of CD44 in response to certain stimulatory events. Transcripts for the CD44 gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full-length nature of some of these variants have not been determined. Alternative splicing is the basis for the structural and functional diversity of the CD44 protein. Diseases associated with CD44 dysfunction include superficial keratitis and lichen sclerosus. CD44 also may be related to tumor metastasis formation.

Especificaciones

Antígeno CD44
Aplicaciones Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence, Western Blot
Clasificación Monoclonal
Clon 6F4H2
Concentración 1.27 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CD44
N.º de referencia del gen P16070
Alias de gen CD44, CD44 antigen, CDW44, CSPG8, ECMR III, Epican, HCELL, Heparan sulfate proteoglycan, Hermes antigen, HUTCH I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU3, MIC4, MUTCH I, PGP 1, PGP I, Pgp1, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Símbolos de los genes CD44
Especie del huésped Mouse
Inmunógeno CD44 Fusion Protein Ag7633
Método de purificación Protein A
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100525150, 960
Especies diana Human, Pig
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.