Learn More
CD44 Mouse anti-Canine, Equine, Alexa Fluor 750, Clone: 69-S5, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems FAB5449S-100UG
Descripción
CD44 Monoclonal specifically detects CD44 in Canine, Equine samples. It is validated for Flow Cytometry.Especificaciones
| CD44 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q28284 | |
| CD44 | |
| Canine marrow cells Accession # Q28284 | |
| RUO | |
| 960 | |
| Canine, Equine | |
| IgG1 |
| Flow Cytometry | |
| 69-S5 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects canine and equine CD44 in Flow Cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
For Research Use Only (RUO)