missing translation for 'onlineSavingsMsg'
Learn More

CD42b Monoclonal Antibody (HIP1), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™

Código de producto. 17880851
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
paquete de 100
paquete de 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17880851 100 Tests paquete de 100
17840851 25 Tests paquete de 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17880851 Proveedor Invitrogen N.º de proveedor H051T03B08

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The HIP1 monoclonal antibody reacts with the human CD42b molecule, a 135 kDa transmembrane protein also known as platelet gpIb. Applications Reported: This HIP1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIP1 antibody has been pre-titrated and tested by flow cytometric analysis of human platelets. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Each NovaFluor conjugate or kit is shipped with CellBlox Blocking Buffer. Use this buffer whenever staining with NovaFluor conjugates, including single color compensation controls using cells. Use 5 μL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells. NovaFluor conjugates are based on Phiton™ technology utilizing novel nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts. Excitation: 509 nm; Emission: 665 nm; Laser: 488 nm (Blue) Laser

CD42b is a platelet activation marker involved in the process of coagulation as an aggregating factor. It interacts with extracellular matrix as well as with adhesion molecules. CD42b expression and activation seems to be regulated by VEGF and PDGF. CD42b composes together with GPIb beta, GPIX and GPV the GPIb-IX-V receptor complex critical in the process of platelet-rich thrombus formation by tethering the platelet to a thrombogenic surface. CD42b binds to von Willebrand factor (VWF) exposed at a site of vascular injury, as well as to thrombin, coagulation factors XI and XII, high molecular wight kininogen, TSP-1, integrin Mac-1 and P-selectin. The extracellular domain of CD42b by its interactions also contributes to metastasis. Further, CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. Several polymorphisms and mutations have been described in the CD42b, some of which are the cause of Bernard-Soulier syndromes and platelet-type von Willebrand disease.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD42b
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon HIP1
Concentración 0.2 μg/Test
Conjugado NovaFluor Blue 660-120S
Formulación PBS with 0.09% sodium azide; pH 7.2
génica Gp1ba
N.º de referencia del gen P07359
Alias de gen Antigen CD42b-alpha; BDPLT1; BDPLT3; BSS; CD42a; CD42B; CD42b-alpha; DBPLT3; Glycocalicin; glycoprotein 1b, alpha polypeptide; glycoprotein Ib (platelet), alpha polypeptide; glycoprotein Ib platelet alpha subunit; glycoprotein Ib platelet subunit alpha; glycoprotein Ibalpha; GP1B; GP1BA; GPIB; GPIb alpha; GP-Ib alpha; GPIbA; GPIbalpha; GPIX; GP-IX; MGC34595; mutant platelet membrane glycoprotein Ib-alpha; platelet glycoprotein Ib alpha chain; platelet glycoprotein Ib alpha polypeptide; Platelet GPIX; platelet membrane glycoprotein 1b-alpha subunit; platelet membrane glycoprotein Ib-alpha; VWDP
Símbolos de los genes Gp1ba
Especie del huésped Mouse
Método de purificación Affinity chromatography
Cantidad 100 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2811
Especies diana Human
Contenido y almacenamiento 4° C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.