missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD41/CD61 Monoclonal Antibody (11C3), PE

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA528368
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute with 1.0 mL distilled water.
CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GPIIb alpha (heavy chain; 120 kDa) and GPIIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
Especificaciones
| CD41/CD61 | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
| 0 | |
| ITGA2B, ITGB3 | |
| Peripheral blood leukocytes from white leghorn chicken. | |
| 100 Tests | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 11C3 | |
| PE | |
| ITGA2B | |
| alphaIIb protein; antigen CD61; BDPLT16; BDPLT2; CD41; CD41B; CD61; glycoprotein GPIIb; glycoprotein GPIIIa; glycoprotein IIb; glycoprotein IIIa; GP2B; GP3A; GPalpha IIb; GPIIb; GPIIIa; GT; GTA; HPA3; INTB3; integrin; integrin alpha 2b; integrin alpha 2b precursor; Integrin alpha-2b; integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; Integrin beta; integrin beta 3; integrin beta 3 subunit; integrin beta3; integrin beta-3; integrin beta-3 subunit; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin subunit beta 3; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41B); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGA2B; ITGA2B-like protein; ITGAB; ITGB3; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/IIIa complex; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet membrane glycoprotein IIb; Platelet membrane glycoprotein IIIa; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 | |
| Mouse | |
| Protein A | |
| RUO | |
| 101749783, 374209 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido