Learn More
CD40 / TNFRSF5 / CD40L-Receptor Mouse, Clone: C40/2383, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 958-MSM3-P0
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| CD40 / TNFRSF5 / CD40L-Receptor | |
| Cell Surface and Secreted | |
| C40/2383 | |
| Unconjugated | |
| Mouse | |
| 43kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 958 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human CD40 protein (apprx. aa 29-107) | |
| L-428 Cells. Tonsil, Spleen or Thymus | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |