Learn More
CD4 Rat, anti-Mouse, Biotin, Clone: RM4-4, STEMCELL Technologies
Rat Monoclonal Antibody
Marca: Stemcell Technologies 60029BT
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The RM4-4 antibody reacts with mouse CD4, an ∼55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobin (Ig) superfamily; CD4 contains four extracellular Ig-like domains
- CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper/inducer cells), and at lower levels on dendritic cells
- In the mouse, CD4 is not expressed by monocytes/macrophages
- CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex
- CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
- Binding of the RM4-4 antibody does not block binding of antibody clones RM4-5 or GK1.5
Especificaciones
| CD4 | |
| Monoclonal | |
| Biotin | |
| 500 μg | |
| Mouse | |
| T Cells, T Cells, CD4+, T Cells, Regulatory |
| Flow Cytometry, Functional Assay | |
| RM4-4 | |
| Rat | |
| Primary | |
| IgG2b κ |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.