missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat, anti-Mouse, APC, Clone: RM4-4, STEMCELL Technologies
Rat Monoclonal Antibody
Marca: Stemcell Technologies 60029AZ.1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The RM4-4 antibody reacts with mouse CD4, an ∼55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobin (Ig) superfamily; CD4 contains four extracellular Ig-like domains
- CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper/inducer cells), and at lower levels on dendritic cells
- In the mouse, CD4 is not expressed by monocytes/macrophages
- CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex
- CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
- Binding of the RM4-4 antibody does not block binding of antibody clones RM4-5 or GK1.5
Especificaciones
| CD4 | |
| Monoclonal | |
| APC | |
| 25 μg | |
| Mouse | |
| T Cells, T Cells, CD4+, T Cells, Regulatory |
| Flow Cytometry, Functional Assay | |
| RM4-4 | |
| Rat | |
| Primary | |
| IgG2b κ |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido