Learn More
CD4 Mouse anti-Human, PerCP-Cy5.5, Clone: SK3 (also known as Leu3a), BD
Descripción
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
Especificaciones
Especificaciones
| Antígeno | CD4 |
| Clasificación | Monoclonal |
| Clon | SK3 (also known as Leu3a) |
| Conjugado | PerCP-Cyanine5.5 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Alias de gen | L3T4, Leu3a, T-cell surface antigen T4/Leu-3, W3/25, CD4 antigen (p55) |
| Especie del huésped | Mouse |
| Inmunógeno | Human Peripheral Blood T Cells |
| Método de purificación | Affinity Purified |
| Cantidad | 25 Tests |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.