missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse, anti-Human, Biotin, Clone: OKT4, STEMCELL Technologies
Descripción
- The OKT4 antibody reacts with CD4, an ∼59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4)
- The epitope for OKT4 has been localized to the D3 domain of the protein, which has a structure resembling an Ig variable domain
- CD4 is expressed at high levels by most thymocytes and a subpopulation of T cells (T-helper cells), and at lower levels by peripheral blood monocytes and macrophages
- Moreover, CD4 is a receptor for human immunodeficiency virus (HIV)
- Binding of the OKT4 antibody to CD4 does not block HIV binding
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD4+ T Cell Isolation Kit (Catalog #19662), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851) and EasySep™ Human CD4 Positive Selection Kit II (Catalog #17852)
Especificaciones
Especificaciones
| Antígeno | CD4 |
| Aplicaciones | Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | OKT4 |
| Conjugado | Biotin |
| Especie del huésped | Mouse |
| Cantidad | 100 μg |
| Primario o secundario | Primary |
| Especies diana | Cynomolgus, Rhesus Monkey, Sooty Mangabey, Chimpanzee, Human |
| Isotype | IgG2b κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?