missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The CD4 antibody from Proteintech is a mouse monoclonal antibody generated with human peripheral t cells of human CD4. This antibody recognizes human antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
Specifications
| Antígeno | CD4 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | OKT4 |
| Concentración | 5 μL (0.125 μg)/Test |
| Conjugado | APC |
| Especie del huésped | Mouse |
| Cantidad | 100 Tests |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 920 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?